Order Entry
Slovenia
ContactUsLinkComponent
Carboxynitroso rubber (CNR)
Carboxynitroso rubber (CNR)
Catalog # APOSPC1672-5G
Supplier:  Apollo Scientific
CAS Number:  
Carboxynitroso rubber (CNR)
Catalog # APOSPC1672-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:25067-94-1
MDL Number:MFCD00166359