Order Entry
Slovenia
ContactUsLinkComponent
2-[(2-Chloropyridin-3-yl)oxy]ethanol 95%
2-[(2-Chloropyridin-3-yl)oxy]ethanol 95%
Catalog # APOSOR959751-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(2-Chloropyridin-3-yl)oxy]ethanol 95%
Catalog # APOSOR959751-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.597 g/mol
Storage Temperature:Ambient
CAS Number:313657-79-3
Formula:C₇H₈ClNO₂
MDL Number:MFCD21120525