Order Entry
Slovenia
ContactUsLinkComponent
3-Methyl-4-isoxazolecarboxylic acid methyl ester 95%
3-Methyl-4-isoxazolecarboxylic acid methyl ester 95%
Catalog # APOSOR955818-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-4-isoxazolecarboxylic acid methyl ester 95%
Catalog # APOSOR955818-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.125 g/mol
Storage Temperature:Ambient
CAS Number:92234-50-9
Formula:C₆H₇NO₃
MDL Number:MFCD12032284