1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:171919-37-2
:
Formula:C₉H₈N₂O₂
MDL Number:MFCD15529044
:176.172 g/mol
Storage Temperature:Ambient
Frequently Bought Together