Order Entry
Slovenia
ContactUsLinkComponent
(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine 97%
(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine 97%
Catalog # APOSOR924457-1G
Supplier:  Apollo Scientific
CAS Number:  
(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine 97%
Catalog # APOSOR924457-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.263 g/mol
Melting Pt:42…44 °C
Storage Temperature:Refrigerator
CAS Number:114676-93-6
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD11109735