(R)-5-Bromomethyl-2-pyrrolidinone 95%
Catalog # APOSOR914328-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.027 g/mol
Storage Temperature:Ambient
CAS Number:98612-60-3
Formula:C₅H₈BrNO
MDL Number:MFCD06799465