Order Entry
Slovenia
ContactUsLinkComponent
Methyl 2-methoxy-3-nitrobenzoate 98%
Irritant
Methyl 2-methoxy-3-nitrobenzoate 98%
Catalog # APOSOR912665-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 2-methoxy-3-nitrobenzoate 98%
Catalog # APOSOR912665-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:48…52 °C
 :211.171 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:90564-26-4
Formula:C₉H₉NO₅
MDL Number:MFCD11045625