2-Methoxy-3-methylaniline 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:18102-30-2
- Formula:C₈H₁₁NO
- MDL Number:MFCD12547890
- Molecular Weight:137.179 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
- CAS Number:18102-30-2
- Formula:C₈H₁₁NO
- MDL Number:MFCD12547890
- Molecular Weight:137.179 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



