Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-4-methylnicotinic acid
6-Chloro-4-methylnicotinic acid
6-Chloro-4-methylnicotinic acid
6-Chloro-4-methylnicotinic acid
APOSOR60150-1G
:
Apollo Scientific
:
6-Chloro-4-methylnicotinic acid
APOSOR60150-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
171.581 g/mol
:
>100 °C
:
GHS07
:
Refrigerator
:
503555-50-8
:
C₇H₆ClNO₂
:
MFCD12755871