Order Entry
Slovenia
ContactUsLinkComponent
Methyl 5-bromo-2-(piperidin-1-ylmethyl)furan-3-carboxylate
Methyl 5-bromo-2-(piperidin-1-ylmethyl)furan-3-carboxylate
Catalog # APOSOR55384-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-bromo-2-(piperidin-1-ylmethyl)furan-3-carboxylate
Catalog # APOSOR55384-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :302.164 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₆BrNO₃
MDL Number:MFCD21364934