Isopropyl 5-chloro-4-methyl-2-nitrobenzoate
Catalog # APOSOR55336-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:257.67 g/mol
Storage Temperature:Ambient
CAS Number:1204518-43-3
Formula:C₁₁H₁₂ClNO₄
MDL Number:MFCD27976845