Methyl 5-hydroxy-2-nitrobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59216-77-2
:
Formula:C₈H₇NO₅
MDL Number:MFCD00100487
:197.145 g/mol
Melting Pt:100…115 °C
Storage Temperature:Ambient
Frequently Bought Together