(8S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:865303-57-7
:
Formula:C₉H₁₂N₂·2ClH
MDL Number:MFCD17926204
:221.127 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together