
Methyl 5-ethynylpyridine-2-carboxylate
Catalog # APOSOR52206-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:161.157 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:17880-61-4
Formula:C₉H₇NO₂
MDL Number:MFCD13189756