3-Bromo-5-nitrobenzyl alcohol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:139194-79-9
:
Formula:C₇H₆BrNO₃
MDL Number:MFCD09056873
:232.031 g/mol
Melting Pt:88…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together