Methyl 4-bromo-6-cyanobenzo[b]thiophene-2-carboxylate ≥98%
Catalog # APOSOR510249-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:296.14 g/mol
Storage Temperature:Ambient
CAS Number:753455-45-7
Formula:C₁₁H₆BrNO₂S