Order Entry
Slovenia
ContactUsLinkComponent
4-Hydroxy-5-methoxy-2-nitrobenzoic acid ≥98%
4-Hydroxy-5-methoxy-2-nitrobenzoic acid ≥98%
Catalog # APOSOR510185-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Hydroxy-5-methoxy-2-nitrobenzoic acid ≥98%
Catalog # APOSOR510185-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :213.144 g/mol
Storage Temperature:Ambient
CAS Number:140647-01-4
Formula:C₈H₇NO₆
MDL Number:MFCD22627819