4-Chloro-2-methyl-5-(methylsulphonyl)benzoic acid ≥98%
Catalog # APOSOR510106-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:248.683 g/mol
Storage Temperature:Ambient
CAS Number:176309-00-5
Formula:C₉H₉ClO₄S
MDL Number:MFCD21099591