Order Entry
Slovenia
ContactUsLinkComponent
 
2-Amino-2-(1-naphthyl)acetic acid 96%
Catalog # APOSOR480441-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-2-(1-naphthyl)acetic acid 96%
Catalog # APOSOR480441-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.221 g/mol
Storage Temperature:Ambient
CAS Number:97611-60-4
Formula:C₁₂H₁₁NO₂
MDL Number:MFCD02662419