1-Methylpiperidine-4-carbaldehyde hydrochloride hydrate 95%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₇H₁₃NO·ClH·H₂O
MDL Number:MFCD28024660
:181.66 g/mol
Boiling Pt:60…62 °C (5 mm Hg)
Density:0.9574 g/cm³ (25 °C)
Storage Temperature:Freezer
UN:1993
ADR:3,III
Frequently Bought Together