3-[Phenyl[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2006278-04-0
:
Formula:C₁₆H₁₇NO₅
MDL Number:MFCD28977378
:303.31 g/mol
Storage Temperature:Ambient
Frequently Bought Together