3-[(3-Pyridyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSOR471396-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:304.298 g/mol
Storage Temperature:Ambient
CAS Number:2006277-53-6
Formula:C₁₅H₁₆N₂O₅
MDL Number:MFCD28977402