Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate
APOSOR470854-1G
:
Apollo Scientific
:
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate
APOSOR470854-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
211.171 g/mol
:
Ambient
:
53732-08-4
:
C₉H₉NO₅
:
MFCD08275104