Methyl 4-morpholino-2-nitrobenzoate
Catalog # APOSOR470502-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:266.25 g/mol
Storage Temperature:Ambient
CAS Number:404010-97-5
Formula:C₁₂H₁₄N₂O₅
MDL Number:MFCD22205882