Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,5-Dichloro-3-nitrobenzoate
Methyl 2,5-Dichloro-3-nitrobenzoate
Methyl 2,5-Dichloro-3-nitrobenzoate
APOSOR470475-5G
:
Apollo Scientific
:
Methyl 2,5-Dichloro-3-nitrobenzoate
APOSOR470475-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
250.036 g/mol
:
Ambient
:
34408-25-8
:
C₈H₅Cl₂NO₄
:
MFCD01529637