4-Methyl-1-piperazinepropionic acid hydrochloride
Catalog # APOSOR470248-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:208.686 g/mol
Storage Temperature:Ambient
CAS Number:775349-40-1
Formula:C₈H₁₆N₂O₂·ClH
MDL Number:MFCD03452741