
3-Iodo-cyclobutanecarboxylic acid ethyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98431-45-9
:
Formula:C₇H₁₁IO₂
MDL Number:MFCD20257322
:254.066 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together