Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Acetyl-3-azetidinecarboxylic acid methyl ester
1-Acetyl-3-azetidinecarboxylic acid methyl ester
1-Acetyl-3-azetidinecarboxylic acid methyl ester
APOSOR451038-5G
:
Apollo Scientific
:
1-Acetyl-3-azetidinecarboxylic acid methyl ester
APOSOR451038-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
157.167 g/mol
:
Ambient
:
1353953-76-0
:
C₇H₁₁NO₃
:
MFCD21098315