
Methyl 3-bromo-5-methylthiophene-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1257535-60-6
:
Formula:C₇H₇BrO₂S
MDL Number:MFCD17168275
:235.098 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together