
Methyl 2-bromo-5-methylthiophene-3-carboxylate
Catalog # APOSOR43540-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:235.098 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:944709-72-2
Formula:C₇H₇BrO₂S
MDL Number:MFCD09702018