(2S,6S)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:574007-66-2
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD11973745
:214.305 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together