2-Methoxy-3-methylbenzaldehyde 95%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:67639-61-6
- Formula:C₉H₁₀O₂
- MDL Number:MFCD07783657
- Molecular Weight:150.175 g/mol
- Melting Pt:44…46 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:67639-61-6
- Formula:C₉H₁₀O₂
- MDL Number:MFCD07783657
- Molecular Weight:150.175 g/mol
- Melting Pt:44…46 °C
- Storage Temperature:Ambient



