
1-Methyl-5-(thien-2-yl)-1H-pyrazole-3-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:869901-15-5
:
Formula:C₉H₈N₂O₂S
MDL Number:MFCD08271952
:208.237 g/mol
Melting Pt:156…158 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together