4-(2-Amino-5-bromo-3-pyridinyl)-2-methyl-3-butyn-2-ol 95%
Catalog # APOSOR370040-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:255.111 g/mol
Storage Temperature:Ambient
CAS Number:1262985-25-0
Formula:C₁₀H₁₁BrN₂O
MDL Number:MFCD19689609