Methyl 6-methylpyrazine-2-carboxylate
Catalog # APOSOR350256-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.151 g/mol
Storage Temperature:Ambient
CAS Number:41110-38-7
Formula:C₇H₈N₂O₂
MDL Number:MFCD08705757