
5-(4-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:91182-89-7
:
Formula:C₁₁H₈ClNO₃
MDL Number:MFCD01571297
:237.639 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together