Methyl 3-hydroxyindole-2-carboxylate
Catalog # APOSOR322584-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:191.183 g/mol
Storage Temperature:Ambient
CAS Number:31827-04-0
Formula:C₁₀H₉NO₃
MDL Number:MFCD00030205