Order Entry
Slovenia
ContactUsLinkComponent
2,2,5,7,8-Pentamethyl-6-chromanol
2,2,5,7,8-Pentamethyl-6-chromanol
Catalog # APOSOR322018-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2,5,7,8-Pentamethyl-6-chromanol
Catalog # APOSOR322018-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.307 g/mol
Storage Temperature:Ambient
CAS Number:950-99-2
Formula:C₁₄H₂₀O₂
MDL Number:MFCD00210347