Order Entry
Slovenia
ContactUsLinkComponent
1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid
Irritant
1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid
Catalog # APOSOR305606-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid
Catalog # APOSOR305606-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.111 g/mol
Melting Pt:161…163 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:50700-55-5
Formula:C₅H₅N₃O₄
MDL Number:MFCD19443277