cis-3-Aminocyclobutanecarbonitrile hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1638765-32-8
:
Formula:C₅H₈N₂·ClH
MDL Number:MFCD28501419
:132.591 g/mol
Storage Temperature:Ambient
Frequently Bought Together