
4-Bromo-1-methyl-1H-indole-6-carboxylic acid
Catalog # APOSOR300192-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:254.08 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1782217-26-8
Formula:C₁₀H₈BrNO₂
MDL Number:MFCD31536723