
3,4-Dimethylthieno[2,3-b]thiophene-2-carboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:159709-36-1
:
Formula:C₉H₈OS₂
MDL Number:MFCD00052282
:196.289 g/mol
Melting Pt:147…149 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together