Order Entry
Slovenia
ContactUsLinkComponent
Methyl 4-bromothiophene-2-carboxylate 98%
Methyl 4-bromothiophene-2-carboxylate 98%
Catalog # APOSOR17259-10G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-bromothiophene-2-carboxylate 98%
Catalog # APOSOR17259-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :221.072 g/mol
Storage Temperature:Refrigerator
CAS Number:62224-16-2
Formula:C₆H₅BrO₂S
MDL Number:MFCD06203668