1-Methyl-1H-indole-7-sulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:941716-95-6
:
Formula:C₉H₈ClNO₂S
MDL Number:MFCD09879958
:229.683 g/mol
Melting Pt:100…103 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together