
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carbaldehyde
Catalog # APOSOR110926-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.213 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:623564-80-7
Formula:C₈H₈O₂S
MDL Number:MFCD18433145