Methyl 8-bromo-6-chloro-2-oxo-2H-chromene-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1427460-59-0
:
Formula:C₁₁H₆BrClO₄
MDL Number:MFCD19442765
:317.52 g/mol
Storage Temperature:Ambient
Frequently Bought Together