Order Entry
Slovenia
ContactUsLinkComponent
Methyl 1-methyl-1H-indole-3-carboxylate
Irritant
Methyl 1-methyl-1H-indole-3-carboxylate
Catalog # APOSOR110449-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 1-methyl-1H-indole-3-carboxylate
Catalog # APOSOR110449-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.211 g/mol
Melting Pt:82…84 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:108438-43-3
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00085570