
2,5-Dimethyl-1-[(pyridin-4-yl)methyl]-1H-pyrrole-3-carboxylic acid 97%
Catalog # APOSOR0416-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:230.262 g/mol
Melting Pt:171…173 °C
:GHS07
Storage Temperature:Ambient
CAS Number:306936-15-2
Formula:C₁₃H₁₄N₂O₂
MDL Number:MFCD01567218