Order Entry
Slovenia
ContactUsLinkComponent
2,4-Dioxo-4-(pyridin-3-yl)butanoic acid 95%
Irritant
2,4-Dioxo-4-(pyridin-3-yl)butanoic acid 95%
Catalog # APOSOR01522-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dioxo-4-(pyridin-3-yl)butanoic acid 95%
Catalog # APOSOR01522-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.156 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:394655-14-2
Formula:C₉H₇NO₄
MDL Number:MFCD06245355