Biotin-[2H2] (Vitamin H)
:1217481-41-8
:
:C₁₀H₁₄D₂N₂O₃S
:MFCD11656103
:246.323 g/mol
:Ambient
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Transfer pipettes
: RATIOLAB
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection